MilliporeSigma
  • Innovative sampling and extraction methods for the determination of nonsteroidal anti-inflammatory drugs in water.
Innovative sampling and extraction methods for the determination of nonsteroidal anti-inflammatory drugs in water.
Journal of pharmaceutical and biomedical analysis (2014-12-01)
Shivani Tanwar, Marina Di Carro, Emanuele Magi
ABSTRACT
Two different innovative approaches were used for the determination of nonsteroidal anti-inflammatory drugs (NSAIDs) in water: stir bar sorptive extraction (SBSE) and passive sampling, followed by electrospray ionization liquid chromatography-tandem mass spectrometry. SBSE was developed by comparing EG-Silicone and PDMS stir bars and optimizing main parameters to attain high preconcentration. Quantitative analysis was carried out by mass spectrometry in negative ionization mode and multiple reaction monitoring. The SBSE-LC-MS/MS method provided satisfactory figures of merit with LOD (7.5-71 ng L(-1)) and LOQ (22.5-213 ng L(-1)). The developed method was successfully applied to real samples collected from river water and wastewater effluents. The obtained results showed the presence of all analytes at trace levels, in a wide range of concentrations. The passive sampling approach was carried out by using Polar Organic Chemical Integrative Sampler (POCIS); samplers were deployed for 15 days in river and tap water, allowing to detect analytes at ultra-trace levels. Time-Weighted Average concentration of NSAIDs in river water was estimated in the range 0.33-0.46 ng L(-1), using the sampling rates previously obtained by means of a simple calibration system.
Product Description
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Mefenamic acid
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
N-Acetyl-D-penicillamine, for HPLC derivatization, ≥99.0% (T)
Supelco
Ibuprofen
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketoprofen-d3, VETRANAL®, analytical standard
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Mefenamic acid, analytical standard
Mefenamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid