MilliporeSigma
  • Targeted intracellular delivery of proteins with spatial and temporal control.
Targeted intracellular delivery of proteins with spatial and temporal control.
Molecular pharmaceutics (2014-12-10)
Demosthenes P Morales, Gary B Braun, Alessia Pallaoro, Renwei Chen, Xiao Huang, Joseph A Zasadzinski, Norbert O Reich
ABSTRACT
While a host of methods exist to deliver genetic materials or small molecules to cells, very few are available for protein delivery to the cytosol. We describe a modular, light-activated nanocarrier that transports proteins into cells by receptor-mediated endocytosis and delivers the cargo to the cytosol by light triggered endosomal escape. The platform is based on hollow gold nanoshells (HGN) with polyhistidine tagged proteins attached through an avidity-enhanced, nickel chelation linking layer; here, we used green fluorescent protein (GFP) as a model deliverable cargo. Endosomal uptake of the GFP loaded nanocarrier was mediated by a C-end Rule (CendR) internalizing peptide fused to the GFP. Focused femtosecond pulsed-laser excitation triggered protein release from the nanocarrier and endosome disruption, and the released protein was capable of targeting the nucleoli, a model intracellular organelle. We further demonstrate the generality of the approach by loading and releasing Sox2 and p53. This method for targeting of individual cells, with resolution similar to microinjection, provides spatial and temporal control over protein delivery.
Product Description
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%