MilliporeSigma
  • Sustained inhibition of HIV-1 replication by conditional expression of the E. coli-derived endoribonuclease MazF in CD4+ T cells.
Sustained inhibition of HIV-1 replication by conditional expression of the E. coli-derived endoribonuclease MazF in CD4+ T cells.
Human gene therapy methods (2013-02-28)
Mika Okamoto, Hideto Chono, Yasuhiro Kawano, Naoki Saito, Hiroshi Tsuda, Koichi Inoue, Ikunoshin Kato, Junichi Mineno, Masanori Baba
ABSTRACT
Gene therapy using a Tat-dependent expression system of MazF, an ACA nucleotide sequence-specific endoribonuclease derived from Escherichia coli, in a retroviral vector appears to be an alternative approach to the treatment of human immunodeficiency virus type 1 (HIV-1) infection. MazF can cleave HIV-1 RNA, since it has more than 240 ACA sequences. Significant inhibition of viral replication, irrespective of HIV-1 strains, was observed in CD4(+) T cells that had been transduced with the MazF-expressing retroviral vector (MazF-T cells). The growth and viability of MazF-T cells were not affected by HIV-1 infection. Interestingly, the infectivity of HIV-1 produced from MazF-T cells was found to be lower than that from control CD4(+) T cells. A long-term culture experiment with HIV-1-infected cells revealed that viral replication was always lower in MazF-T cells than in CD4(+) T cells transduced with or without a control vector for more than 200 days. MazF was expressed and mainly localized in the cytoplasm of the infected cells. Unlike in CD4(+) T cells, the expression level of Tat gradually decreased rather than increased in MazF-T cells after HIV-1 infection. As a consequence, the expression level of MazF appeared to be well regulated and sustained during HIV-1 infection in MazF-T cells. Furthermore, the levels of cellular mRNA were not affected by HIV-1 infection. Thus, the Tat-dependent MazF expression system has great potential for inhibition of HIV-1 replication in vivo without apparent toxicity and may be able to avoid the emergence of resistant strains.
Sigma-Aldrich
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Human IFN γ ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IFN γ ELISA Kit, for serum, plasma, cell culture supernatant and urine