MilliporeSigma
  • A nonsteroidal glucocorticoid receptor agonist inhibits allergen-induced late asthmatic responses.
A nonsteroidal glucocorticoid receptor agonist inhibits allergen-induced late asthmatic responses.
American journal of respiratory and critical care medicine (2014-12-05)
Gail M Gauvreau, Louis-Philippe Boulet, Richard Leigh, Donald W Cockcroft, Kieran J Killian, Beth E Davis, Francine Deschesnes, Richard M Watson, Veronica Swystun, Carina Kärrman Mårdh, Peter Wessman, Carin Jorup, Magnus Aurivillius, Paul M O'Byrne
ABSTRACT
Effective antiinflammatory therapies are needed for the treatment of asthma, but preferably without the systemic adverse effects of glucocorticosteroids. We evaluated the effect of an inhaled nonsteroidal glucocorticoid receptor agonist, AZD5423, on allergen-induced responses. Twenty subjects with mild allergic asthma were randomized to receive 7 days of treatment with nebulized AZD5423 (75 or 300 μg) once daily, budesonide 200 μg twice daily via Turbuhaler, or placebo in a double-blind, four-period, crossover design study. Allergen challenge was performed on Day 6. FEV1 was measured repeatedly for 7 hours after allergen challenge for early and late asthmatic responses. Sputum inflammatory cells was measured before and at 7 and 24 hours after allergen challenge, and methacholine airway responsiveness was measured before and 24 hours after allergen challenge. AZD5423 significantly attenuated the fall in FEV1 during the late asthmatic response (both doses led to an 8.7% fall) versus placebo (14% fall) (P < 0.05) with no effect of budesonide (12.5% fall) versus placebo (P > 0.05). There was no effect on the fall in FEV1 during early asthmatic response. AZD5423 300 and 75 μg significantly attenuated allergen-induced sputum eosinophilia by 63 and 61% at 7 hours, respectively, and by 46 and 34% at 24 hours after allergen challenge, respectively, versus placebo (all P < 0.05). Budesonide did not reduce allergen-induced sputum eosinophilia versus placebo. AZD5423 at 300 μg significantly attenuated allergen-induced airway hyperresponsiveness at 24 hours after allergen challenge versus placebo (P < 0.05). Both doses of AZD5423 were well tolerated. Seven-day treatment with inhalation of the nonsteroidal glucocorticoid receptor agonist AZD5423 effectively reduced allergen-induced responses in subjects with mild allergic asthma. Clinical trial registered with www.clinicaltrials.gov (NCT01225549).
Product Description
Supelco
Budesonide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Budesonide, ≥99%
Budesonide, European Pharmacopoeia (EP) Reference Standard