MilliporeSigma
  • Application of hygrine and cuscohygrine as possible markers to distinguish coca chewing from cocaine abuse on WDT and forensic cases.
Application of hygrine and cuscohygrine as possible markers to distinguish coca chewing from cocaine abuse on WDT and forensic cases.
Forensic science international (2014-03-25)
N C Rubio, S Strano-Rossi, M J Tabernero, J L Gonzalez, L Anzillotti, M Chiarotti, A M Bermejo
ABSTRACT
The objectives of present work are twofold. First, we want to verify that hygrine and cuscohygrine are good markers to distinguish between chewing coca leaves and cocaine abuse. Secondly, we try to develop a quick and easy qualitative method to determine the two mentioned markers. We analyzed two kinds of urine samples: the first group consisted of twenty-four (24) subjects: urine samples were obtained from various types of workers (e.g. doctors, chemists, nurses, technicians, painters, contractors, employees and some retired persons) who admitted chewing coca leaves. Frequency of the habit of chewing coca leaves was variable. They practiced "coqueo" between two (2) and forty-four (44) years. Sixteen (16) of them used alkaline substances to enhance the extraction of cocaine from the leaves The second group of urine samples consisted on thirty-eight (38) cocaine abusers, from forensic cases from Spain and Argentina. A GC/MS qualitative method, performed after liquid-liquid extraction, was developed and validated (the parameters studied were selectivity/specificity, LOD and stability), and then applied to the urine samples. Hygrine and cuscohygrine are good markers to distinguish between chewing coca leaves and cocaine abuse, and the qualitative method presented can be used successfully in workplace drug testing and forensic cases.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%