MilliporeSigma
  • Morphological, molecular, and biochemical characterization of astaxanthin-producing green microalga Haematococcus sp. KORDI03 (Haematococcaceae, Chlorophyta) isolated from Korea.
Morphological, molecular, and biochemical characterization of astaxanthin-producing green microalga Haematococcus sp. KORDI03 (Haematococcaceae, Chlorophyta) isolated from Korea.
Journal of microbiology and biotechnology (2014-11-11)
Ji Hyung Kim, Abu Affan, Jiyi Jang, Mee-Hye Kang, Ah-Ra Ko, Seon-Mi Jeon, Chulhong Oh, Soo-Jin Heo, Youn-Ho Lee, Se-Jong Ju, Do-Hyung Kang
ABSTRACT
A unicellular red microalga was isolated from environmental freshwater in Korea, and its morphological, molecular, and biochemical properties were characterized. Morphological analysis revealed that the isolate was a unicellular biflagellated green microalga that formed a non-motile, thick-walled palmelloid or red aplanospore. To determine the taxonomical position of the isolate, its 18S rRNA and rbcL genes were sequenced and phylogenetic analysis was performed. We found that the isolate was clustered together with other related Haematococcus strains showing differences in the rbcL gene. Therefore, the isolated microalga was classified into the genus Haematococcus, and finally designated Haematococcus sp. KORDI03. The microalga could be cultivated in various culture media under a broad range of pH and temperature conditions. Compositions of the microalgal cellular components were analyzed, and its protein, carbohydrate, and lipid compositions were estimated to be 21.1 ± 0.2%, 48.8 ± 1.8%, and 22.2 ± 0.9%, respectively. In addition, D-glucose and D-mannose were the dominant monosaccharides in the isolate, and its amino acids were composed mainly of aspartic acid, glutamic acid, alanine, and leucine. Moreover, several polyunsaturated fatty acids accounted for about 80% of the total fatty acids in Haematococcus sp. KORDI03, and the astaxanthin content in the red aplanospores was estimated to be 1.8% of the dry cell weight. To the best of our knowledge, this is the first report of an Haematococcus sp. isolated from Korea, which may be used for bioresource production in the microalgal industry.
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Astaxanthin, ≥97% (HPLC)
Supelco
all-trans-Astaxanthin, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Nonadecanoic acid, ≥98% (GC)
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Nonadecanoic acid, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%