MilliporeSigma
  • Frequency of CD4+CXCR5+ TFH cells in patients with hepatitis b virus-associated membranous nephropathy.
Frequency of CD4+CXCR5+ TFH cells in patients with hepatitis b virus-associated membranous nephropathy.
International immunopharmacology (2014-07-01)
Yong Liu, Pingwei Zhao, Zhihui Qu, Desalegn Admassu Ayana, Yanfang Jiang
ABSTRACT
The frequency of different subsets of CD4(+)CXCR5(+) TFH cells and serum cytokine levels were analyzed in a total of 14 patients with newly diagnosed hepatitis B virus-associated membranous nephropathy (HBV-MN), 12 individuals with immune-tolerant HBV infection (HBV-IT) and 12 healthy controls (HC). Serum cytokine levels were measured before and 10-12 weeks after treatment. Significantly higher frequency of CD4(+)CXCR5(+), CD4(+)CXCR5(+)ICOS(+) and CD4(+)CXCR5(+)PD-1(+) TFH cells, and higher serum levels of IL-17A, IFN-γ, IL-2, IL-10, IL-4 and IL-21 were detected in HBV-MN patients compared to the HC. The percentage of CD4(+)CXCR5(+) TFH cells and serum IL-21 level in HBV-MN patients were also higher than the HBV-IT. The percentage of CD4(+)CXCR5(+) TFH cell was negatively correlated with the value of eGFR, and the percentage of CD4(+)CXCR5(+)ICOS(+) TFH cells was positively correlated with the 24-h urinary protein concentration. Notably, the percentage of CD4(+)CXCR5(+)PD-1(+) TFH cells was positively correlated with serum IL-21 level and 24-h urinary protein concentration. Treatment with prednisone or/and immunosuppressive drugs significantly reduced the frequency of CD4(+)CXCR5(+), CD4(+)CXCR5(+)PD-1(+) TFH cells and serum IL-21 level, but increased IL-4 and IL-10 levels in the patients. CD4(+)CXCR5(+) TFH cells, especially CD4(+)CXCR5(+)PD-1(+) TFH cells may participate in the pathogenesis of HBV-MN.
Sigma-Aldrich
Human IL21 / Interleukin-21 ELISA Kit