MilliporeSigma
  • Functional characterization of wild-type and 49 CYP2D6 allelic variants for N-desmethyltamoxifen 4-hydroxylation activity.
Functional characterization of wild-type and 49 CYP2D6 allelic variants for N-desmethyltamoxifen 4-hydroxylation activity.
Drug metabolism and pharmacokinetics (2014-03-22)
Yuka Muroi, Takahiro Saito, Masamitsu Takahashi, Kanako Sakuyama, Yui Niinuma, Miyabi Ito, Chiharu Tsukada, Kiminori Ohta, Yasuyuki Endo, Akifumi Oda, Noriyasu Hirasawa, Masahiro Hiratsuka
ABSTRACT
Genetic variations in cytochrome P450 2D6 (CYP2D6) contribute to interindividual variability in the metabolism of clinically used drugs, e.g., tamoxifen. CYP2D6 is genetically polymorphic and is associated with large interindividual variations in therapeutic efficacy and drug toxicity. In this study, we performed an in vitro analysis of 50 allelic variants of CYP2D6 proteins. Wild-type CYP2D6.1 and 49 variants were transiently expressed in COS-7 cells, and the enzymatic activities of the CYP2D6 variants were characterized using N-desmethyltamoxifen as a substrate. The kinetic parameters K(m), V(max), and intrinsic clearance (V(max)/K(m)) of N-desmethyltamoxifen 4-hydroxylation were determined. Among the 50 CYP2D6 variants, the kinetic parameters for N-desmethyltamoxifen 4-hydroxylation were determined for 20 CYP2D6 variants. On the other hand, the kinetic parameters of 30 CYP2D6 variants could not be determined because the amount of metabolite produced was at or below the detection limit at the lower substrate concentrations. Among them, 8 variants, i.e., CYP2D6.2, .9, .26, .28, .32, .43, .45, and .70, showed decreased intrinsic clearance at <50% of CYP2D6.1. The comprehensive in vitro assessment of CYP2D6 variants provides novel insights into allele-specific activity towards tamoxifen and may be valuable when interpreting in vivo studies.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Supelco
(±)-Propranolol hydrochloride, analytical standard
USP
Dextromethorphan, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard