MilliporeSigma
  • DNA polymorphism and epigenetic marks modulate the affinity of a scaffold/matrix attachment region to the nuclear matrix.
DNA polymorphism and epigenetic marks modulate the affinity of a scaffold/matrix attachment region to the nuclear matrix.
European journal of human genetics : EJHG (2014-01-23)
Natalia P Kisseljova, Petr Dmitriev, Alexey Katargin, Elena Kim, Daria Ezerina, Diana Markozashvili, Daria Malysheva, Emmeline Planche, Richard J L F Lemmers, Silvère M van der Maarel, Dalila Laoudj-Chenivesse, Marc Lipinski, Yegor S Vassetzky
ABSTRACT
Mechanisms that regulate attachment of the scaffold/matrix attachment regions (S/MARs) to the nuclear matrix remain largely unknown. We have studied the effect of simple sequence length polymorphism (SSLP), DNA methylation and chromatin organization in an S/MAR implicated in facioscapulohumeral dystrophy (FSHD), a hereditary disease linked to a partial deletion of the D4Z4 repeat array on chromosome 4q. This FSHD-related nuclear matrix attachment region (FR-MAR) loses its efficiency in myoblasts from FSHD patients. Three criteria were found to be important for high-affinity interaction between the FR-MAR and the nuclear matrix: the presence of a specific SSLP haplotype in chromosomal DNA, the methylation of one specific CpG within the FR-MAR and the absence of histone H3 acetylated on lysine 9 in the relevant chromatin fragment.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Anti-TFCP2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Cytosine, ≥99%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard