MilliporeSigma
  • Retinoic acid primes human dendritic cells to induce gut-homing, IL-10-producing regulatory T cells.
Retinoic acid primes human dendritic cells to induce gut-homing, IL-10-producing regulatory T cells.
Mucosal immunology (2014-07-17)
G Bakdash, L T C Vogelpoel, T M M van Capel, M L Kapsenberg, E C de Jong
ABSTRACT
The vitamin A metabolite all-trans retinoic acid (RA) is an important determinant of intestinal immunity. RA primes dendritic cells (DCs) to express CD103 and produce RA themselves, which induces the gut-homing receptors α4β7 and CCR9 on T cells and amplifies transforming growth factor (TGF)-β-mediated development of Foxp3(+) regulatory T (Treg) cells. Here we investigated the effect of RA on human DCs and subsequent development of T cells. We report a novel role of RA in immune regulation by showing that RA-conditioned human DCs did not substantially enhance Foxp3 but induced α4β7(+) CCR9(+) T cells expressing high levels of interleukin (IL)-10, which were functional suppressive Treg cells. IL-10 production was dependent on DC-derived RA and was maintained when DCs were stimulated with toll-like receptor ligands. Furthermore, the presence of TGF-β during RA-DC-driven T-cell priming favored the induction of Foxp3(+) Treg cells over IL-10(+) Treg cells. Experiments with naive CD4(+) T cells stimulated by anti-CD3 and anti-CD28 antibodies in the absence of DCs emphasized that RA induces IL-10 in face of inflammatory mediators. The data thus show for the first time that RA induces IL-10-producing Treg cells and postulates a novel mechanism for IL-10 in maintaining tolerance to the intestinal microbiome.
Product Description
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)