MilliporeSigma
  • People, pets, and parasites: one health surveillance in southeastern Saskatchewan.
People, pets, and parasites: one health surveillance in southeastern Saskatchewan.
The American journal of tropical medicine and hygiene (2014-03-19)
Janna M Schurer, Momar Ndao, Helen Quewezance, Stacey A Elmore, Emily J Jenkins
ABSTRACT
Residents of remote and Indigenous communities might experience higher exposure to some zoonotic parasites than the general North American population. Human sero-surveillance conducted in two Saulteaux communities found 113 volunteers exposed as follows: Trichinella (2.7%), Toxocara canis (4.4%), Echinococcus (4.4%), and Toxoplasma gondii (1.8%). In dogs, 41% of 51 fecal samples were positive for at least one intestinal parasite, 3% of 77 were sero-positive for Borrelia burgdorferi, and 21% of 78 for T. gondii. Echinococcus exposure was more likely to occur in non-dog owners (odds ratio [OR]: 11.4, 95% confidence interval [CI]: 1.2-107, P = 0.03); while T. canis was more likely to occur in children (ages 4-17) (OR: 49, 95% CI: 3.9-624; P = 0.003), and those with a history of dog bites (OR: 13.5, 95% CI: 1.02-179; P = 0.048). Our results emphasize the use of dogs as sentinels for emerging pathogens such as Lyme disease, and the need for targeted surveillance and intervention programs tailored for parasite species, cultural groups, and communities.
Product Description
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sucrose, European Pharmacopoeia (EP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture