MilliporeSigma
  • Astaxanthin activates nuclear factor erythroid-related factor 2 and the antioxidant responsive element (Nrf2-ARE) pathway in the brain after subarachnoid hemorrhage in rats and attenuates early brain injury.
Astaxanthin activates nuclear factor erythroid-related factor 2 and the antioxidant responsive element (Nrf2-ARE) pathway in the brain after subarachnoid hemorrhage in rats and attenuates early brain injury.
Marine drugs (2014-12-23)
Qi Wu, Xiang-Sheng Zhang, Han-Dong Wang, Xin Zhang, Qing Yu, Wei Li, Meng-Liang Zhou, Xiao-Liang Wang
ABSTRACT
Astaxanthin (ATX) has been proven to ameliorate early brain injury (EBI) after experimental subarachnoid hemorrhage (SAH) by modulating cerebral oxidative stress. This study was performed to assess the effect of ATX on the Nrf2-ARE pathway and to explore the underlying molecular mechanisms of antioxidant properties of ATX in EBI after SAH. A total of 96 male SD rats were randomly divided into four groups. Autologous blood was injected into the prechiasmatic cistern of the rat to induce an experimental SAH model. Rats in each group were sacrificed at 24 h after SAH. Expressions of Nrf2 and heme oxygenase-1 (HO-1) were measured by Western blot and immunohistochemistry analysis. The mRNA levels of HO-1, NAD (P) H: quinone oxidoreductase 1 (NQO-1), and glutathione S-transferase-α1 (GST-α1) were determined by real-time polymerase chain reaction (PCR). It was observed that administration of ATX post-SAH could up-regulate the cortical expression of these agents, mediated in the Nrf2-ARE pathway at both pretranscriptional and posttranscriptional levels. Meanwhile, oxidative damage was reduced. Furthermore, ATX treatment significantly attenuated brain edema, blood-brain barrier (BBB) disruption, cellular apoptosis, and neurological dysfunction in SAH models. This study demonstrated that ATX treatment alleviated EBI in SAH model, possibly through activating the Nrf2-ARE pathway by inducing antioxidant and detoxifying enzymes.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard