MilliporeSigma
  • Simultaneous determination of bioactive components of Radix Angelicae Sinensis-Radix Paeoniae Alba herb couple in rat plasma and tissues by UPLC-MS/MS and its application to pharmacokinetics and tissue distribution.
Simultaneous determination of bioactive components of Radix Angelicae Sinensis-Radix Paeoniae Alba herb couple in rat plasma and tissues by UPLC-MS/MS and its application to pharmacokinetics and tissue distribution.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-14)
Niancui Luo, Zhenhao Li, Dawei Qian, Yefei Qian, Jianming Guo, Jin-Ao Duan, Min Zhu
ABSTRACT
A highly sensitive and rapid ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) has been developed and validated for simultaneous quantification of seven components in rat plasma and five components in rat tissues after oral administration of the extracts of different combination Radix Angelicae Sinensis-Radix Paeoniae Alba herb couple and has been applied to compare the different pharmacokinetics and tissue distribution properties of these bioactive components. The extracts of Radix Angelicae Sinensis (RAS), Radix Paeoniae Alba (RPA) and Radix Angelicae Sinensis-Radix Paeoniae Alba herb couple (RRHC) were orally administrated to rats, respectively. The concentrations of ferulic acid, caffeic acid, vanillic acid, ligustilide, paeoniflorin, albiflorin and oxypaeoniflorin in rat plasma and the concentrations of ferulic acid, vanillic acid, paeoniflorin, albiflorin and oxypaeoniflorin in tissues were determined by UPLC-MS/MS. The plasma samples were pretreated by protein precipitation with methanol and the tissue samples were homogenated with water and pretreated by protein precipitation with methanol. Chromatographic separation was performed on a C18 column using 0.1% formic acid-acetonitrile as mobile phase for gradient elution. A triple quadrupole (TQ) tandem mass spectrometry equipped with an electrospray ionization source was used as detector operating both in positive and negative ionization mode and operated by multiple-reaction monitoring (MRM) scanning. Noncompartmental pharmacokinetic parameters were calculated by DAS 2.0 program. The differences between each group were compared by SPSS 16.0 with Independent-Samples T-test. The pharmacokinetic parameters (such as Cmax, Tmax, T1/2, AUC0-T, MRT0-T, Vz/F or CLz/F) of all the detected components between the single herb (RAS or RPA) and herb pair (RRHP) showed significant differences (P<0.05). It indicated that the compatibility of RAS and RPA could alter the pharmacokinetics features of each component. Tissue distribution results showed that ferulic acid, vanillic acid, paeoniflorin, albiflorin and oxypaeoniflorin mostly distributed in liver and kidney both in herb couple and single herb distributed most in liver and kidney. Compared with single herb, RRHC could increase or decrease the concentrations of five components at different time points compared with the sing herb. The results indicated the method was successfully applied to the comparative study on pharmacokinetics and tissue distribution of different combination of RRHC in rats. The compatibility of two Chinese herbs could alter the pharmacokinetics and tissue distribution properties of major bio-active components in the single herb. The results might be helpful for further investigation of compatibility mechanism of RRHC.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%