MilliporeSigma
  • Hyperbaric oxygen therapy increases insulin sensitivity in overweight men with and without type 2 diabetes.
Hyperbaric oxygen therapy increases insulin sensitivity in overweight men with and without type 2 diabetes.
Diving and hyperbaric medicine (2015-05-13)
David Wilkinson, Mirjam Nolting, Mohd Kaisan Mahadi, Ian Chapman, Leonie Heilbronn
ABSTRACT
The onset of insulin resistance is an important metabolic event in the development of type 2 diabetes. For patients with type 2 diabetes, we recently showed that peripheral insulin sensitivity was increased during hyperbaric oxygen treatment (HBOT). This study aims to investigate whether this occurs in a non-patient population with and without type 2 diabetes, along with the mechanism of this effect. Overweight and obese male participants were recruited from the community, 11 without and eight with type 2 diabetes. Insulin sensitivity was measured by the glucose infusion rate (GIR) during a hyperinsulinaemic euglycaemic clamp (80 mU·m⁻²·min⁻¹) at baseline and during the third HBOT session. Monocyte chemo-attractant protein-1 (MCP-1), tumour necrosis factor-α (TNF-α) and interleukin-6 (IL-6) were measured in fasting serum and adipose tissue samples taken for their gene expression at baseline and immediately following four HBOT sessions. Additional fasting serum samples were collected during the first HBOT at 0, 60 and 120 minutes, and 24-hours after the last HBOT. In response to HBOT, GIR was increased by 29±32% in those without (n=10, P=0.01), and by 57±66% in those with type 2 diabetes (n=7, P=0.04). This increase was maintained for 30 minutes post HBOT. Reduced MCP-1 and TNF-α were observed after HBOT, whereas IL-6 was increased only in individuals without diabetes and this correlated with the increase in insulin sensitivity (r²=0.72, P=0.004). Peripheral insulin sensitivity was increased following HBOT in overweight or obese males with and without type 2 diabetes; this increase was maintained for at least 30 minutes post HBOT. Changes in inflammatory cytokines may partly explain this effect.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Supelco
D-(+)-Glucose, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
$52.10
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Please contact Customer Service for Availability
$99.40
1 kg
Please contact Customer Service for Availability
$242.00
5 kg
Please contact Customer Service for Availability
$949.00
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$47.70
1 kg
In Stock
Details...
$54.00
5 kg
In Stock
Details...
$164.00
10 kg
In Stock
Details...
$243.00
25 kg
In Stock
Details...
$473.00
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Please contact Customer Service for Availability
$51.20
500 g
Please contact Customer Service for Availability
$64.60
5 kg
Please contact Customer Service for Availability
$383.00
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$55.10
1 kg
In Stock
Details...
$66.60
5 kg
In Stock
Details...
$206.00
10 kg
Estimated to ship on March 13, 2025
Details...
$287.00
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
Please contact Customer Service for Availability
$70.80
10 kg
Please contact Customer Service for Availability
$288.00
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Please contact Customer Service for Availability
$82.10
1 kg
Please contact Customer Service for Availability
$222.00
5 kg
Please contact Customer Service for Availability
$887.00
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
Please contact Customer Service for Availability
$67.10
500 g
Please contact Customer Service for Availability
$90.90
1 kg
Please contact Customer Service for Availability
$117.00
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
$683.00
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
In Stock
Details...
$103.00
500 g
In Stock
Details...
$157.00
1 kg
In Stock
Details...
$222.00
2.5 kg
In Stock
Details...
$516.00
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material