MilliporeSigma
  • Spp24 derivatives stimulate a Gi-protein coupled receptor-Erk1/2 signaling pathway and modulate gene expressions in W-20-17 cells.
Spp24 derivatives stimulate a Gi-protein coupled receptor-Erk1/2 signaling pathway and modulate gene expressions in W-20-17 cells.
Journal of cellular biochemistry (2014-12-17)
Ke-Wei Zhao, Elsa J Brochmann Murray, Samuel S Murray
ABSTRACT
Secreted phosphoprotein 24 kDa (Spp24) is an apatite- and BMP/TGF-β cytokine-binding phosphoprotein found in serum and many tissues, including bone. N-terminally intact degradation products ranging in size from 14 kDa to 23 kDa have been found in bone. The cleavage sites in Spp24 that produce these short forms have not been definitively identified, and the biological activities and mechanisms of action of Spp24 and its degradation products have not been fully elucidated. We found that the C-terminus of Spp24 is labile to proteolysis by furin, kallikrein, lactoferrin, and trypsin, indicating that both extracellular and intracellular proteolytic events could account for the generation of biologically-active Spp18, Spp16, and Spp14. We determined the effects of these truncation products on kinase-mediated signal transduction, gene expression, and osteoblastic differentiation in W-20-17 bone marrow stromal cells cultured in basal or pro-osteogenic media. After culturing for five days, all forms inhibited BMP-2-stimulated osteoblastic differentiation, assessed as induction of alkaline phosphatase activity, in basal, but not pro-osteogenic media. After 10 days, they also inhibited BMP-2-stimulated mineral deposition in pro-osteogenic media. Spp24 had no effect on Erk1/2 phosphorylation, but Spp18 stimulated short-term Erk1/2, MEK 1/2, and p38 phosphorylation. Pertussis toxin and a MEK1/2 inhibitor ablated Spp18-stimulated Erk 1/2 phosphorylation, indicating a role for Gi proteins and MEK1/2 in the Spp18-stimulated Erk1/2 phosphorylation cascade. Truncation products, but not full-length Spp24, stimulated RUNX2, ATF4, and CSF1 transcription. This suggests that Spp24 truncation products have effects on osteoblastic differentiation mediated by kinase pathways that are independent of exogenous BMP/TGF-β cytokines.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Furin human, ≥2,000 unit/mL, buffered aqueous solution, recombinant, expressed in baculovirus infected Sf9 cells
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Fenbendazole impurity A, European Pharmacopoeia (EP) Reference Standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Carbendazim, PESTANAL®, analytical standard
Sigma-Aldrich
Carbendazim, 97%