MilliporeSigma
  • Fabrication of three-dimensionally interconnected nanoparticle superlattices and their lithium-ion storage properties.
Fabrication of three-dimensionally interconnected nanoparticle superlattices and their lithium-ion storage properties.
Nature communications (2015-03-06)
Yucong Jiao, Dandan Han, Yi Ding, Xianfeng Zhang, Guannan Guo, Jianhua Hu, Dong Yang, Angang Dong
ABSTRACT
Three-dimensional superlattices consisting of nanoparticles represent a new class of condensed materials with collective properties arising from coupling interactions between close-packed nanoparticles. Despite recent advances in self-assembly of nanoparticle superlattices, the constituent materials have been limited to those that are attainable as monodisperse nanoparticles. In addition, self-assembled nanoparticle superlattices are generally weakly coupled due to the surface-coating ligands. Here we report the fabrication of three-dimensionally interconnected nanoparticle superlattices with face-centered cubic symmetry without the presynthesis of the constituent nanoparticles. We show that mesoporous carbon frameworks derived from self-assembled supercrystals can be used as a robust matrix for the growth of nanoparticle superlattices with diverse compositions. The resulting interconnected nanoparticle superlattices embedded in a carbon matrix are particularly suitable for energy storage applications. We demonstrate this by incorporating tin oxide nanoparticle superlattices as anode materials for lithium-ion batteries, and the resulting electrochemical performance is attributable to their unique architectures.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Supelco
Oleic acid, Selectophore, ≥99%