MilliporeSigma
  • Methylcobalamin--a form of vitamin B12 identified and characterised in Chlorella vulgaris.
Methylcobalamin--a form of vitamin B12 identified and characterised in Chlorella vulgaris.
Food chemistry (2014-10-13)
Anantharajappa Kumudha, Sagaya Selvakumar, Pullancheri Dilshad, Gopal Vaidyanathan, Munna Singh Thakur, Ravi Sarada
ABSTRACT
Vitamin B12 is among the most essential biomolecules required for crucial metabolic processes in humans. Vitamin B12 was extracted from Chlorella vulgaris biomass under aqueous conditions, partially purified by passing the extract through amberlite XAD-2, Sep-Pak columns, and further purified by HPLC. The target peak eluent was subjected to characterisation by tandem mass spectrometry (MS/MS), selected ion recording (SIR) and multiple reaction monitoring (MRM) and identified as methylcobalamin (Me-Cbl). Quantification of Me-Cbl was carried out by microbiological and chemiluminescence methods, and found to be 29.87±2 μg/100 g and 26.84±2 μg/100 g dry weight, respectively. The presence of Me-Cbl was further substantiated using gold nanoparticle (AuNPs) based aptamer analysis, and found to be 28.02±2 μg/100 g dry weight. Good similarity was observed among all the methods. Methylcobalamin, a form of vitamin B12 was identified in C. vulgaris and this finding enhances its use as a nutritional supplement.
Product Description
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Urea hydrogen peroxide, 97%
Sigma-Aldrich
Hydrogen peroxide–Urea adduct, powder, 15-17% active oxygen basis
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Methylcobalamin, vitamin B12 analog
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Luminol, ≥97% (HPLC)