MilliporeSigma
  • RFX1-dependent activation of SHP-1 induces autophagy by a novel obatoclax derivative in hepatocellular carcinoma cells.
RFX1-dependent activation of SHP-1 induces autophagy by a novel obatoclax derivative in hepatocellular carcinoma cells.
Oncotarget (2014-06-24)
Jung-Chen Su, Ping-Hui Tseng, Cheng-Yi Hsu, Wei-Tien Tai, Jui-Wen Huang, Ching-Huai Ko, Mai-Wei Lin, Chun-Yu Liu, Kuen-Feng Chen, Chung-Wai Shiau
ABSTRACT
Obatoclax is a small molecule which targets the Bcl-2 family, and is to treat leukemia, lymphoma and lung carcinoma. Previously, an obatoclax analogue, SC-2001, was found to disrupt the protein-protein interactions of the Bcl-2 family and also repress Bcl-XL and Mcl-1 expression via STAT3 inactivation. Here, we report a novel mechanism of autophagy induction by SC-2001 in liver cancer cells. The findings indicate that SC-2001 induced the autophagy marker LC3-II in four hepatocellular carcinoma (HCC) cells. Autophagosomes induced by SC-2001-treated cells were confirmed by electron microscopy. SC-2001 activated SHP-1, dephosphorylated STAT3 and Mcl-1, and subsequently released free beclin 1. Overexpression of STAT3 and Mcl-1 in PLC5 cells attenuated the induction of SC-2001 on autophagy. Abolishment of SHP-1 by a specific inhibitor aboragated the autophagic effects induced by SC-2001. In addition, it was further revealed that RFX-1, a transcription factor of SHP-1, is a critical regulator in SC-2001-mediated autophagy. Downregulation of RFX-1 by si-RNA protected cells from SC-2001-induced autophagy. Importantly, Huh7 tumor-bearing nude mice treated with SC-2001 showed downregulation of Mcl-1 and p-STAT3 protein expression and upregulation of SHP-1, LC3II, and RFX-1 protein expression. In summary, our data suggest that SC-2001 induces autophagic cell death in a RFX1/SHP-1/STAT3/Mcl-1 signaling cascade.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free