MilliporeSigma
  • Discovery of protein disulfide isomerase P5 inhibitors that reduce the secretion of MICA from cancer cells.
Discovery of protein disulfide isomerase P5 inhibitors that reduce the secretion of MICA from cancer cells.
Chembiochem : a European journal of chemical biology (2014-06-13)
Tomohisa Horibe, Aya Torisawa, Yukiko Okuno, Koji Kawakami
ABSTRACT
In order to regulate the activity of P5, which is a member of the protein disulfide isomerase family, we screened a chemical compound library for P5-specific inhibitors, and identified two candidate compounds (anacardic acid and NSC74859). Interestingly, anacardic acid inhibited the reductase activity of P5, but did not inhibit the activity of protein disulfide isomerase (PDI), thiol-disulfide oxidoreductase ERp57, or thioredoxin. NSC74859 inhibited all these enzymes. When we examined the effects of these compounds on the secretion of soluble major histocompatibility complex class-I-related gene A (MICA) from cancer cells, anacardic acid was found to decrease secretion. In addition, anacardic acid was found to reduce the concentration of glutathione up-regulated by the anticancer drug 17-demethoxygeldanamycin in cancer cells. These results suggest that anacardic acid can both inhibit P5 reductase activity and decrease the secretion of soluble MICA from cancer cells. It might be a novel and potent anticancer treatment by targeting P5 on the surface of cancer cells.
Sigma-Aldrich
Anacardic acid
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Human MICA ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2-Hydroxy-4-methoxybenzoic acid, 99%