MilliporeSigma
Hippocampal--prefrontal BDNF and memory for fear extinction.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2014-03-15)
Luis E Rosas-Vidal, Fabricio H Do-Monte, Francisco Sotres-Bayon, Gregory J Quirk
ABSTRACT
Infusing brain-derived neurotrophic factor (BDNF) into the infralimbic (IL) prefrontal cortex is capable of inducing extinction. Little is known, however, about the circuits mediating BDNF effects on extinction or the extent to which extinction requires BDNF in IL. Using local pharmacological infusion of BDNF protein, or an antibody against BDNF, we found that BDNF in the IL, but not prelimbic (PL) prefrontal cortex, is both necessary and sufficient for fear extinction. Furthermore, we report that BDNF in IL can induce extinction of older fear memories (14 days) as well as recent fear memories (1 day). Using immunocytochemistry, we show that BDNF is increased in the ventral hippocampus (vHPC), but not IL or PL, following extinction training. Finally, we observed that infusing BDNF into the vHPC increased the firing rate of IL, but not PL neurons in fear conditioned rats. These findings indicate that an extinction-induced increase in BDNF within the vHPC enhances excitability in IL targets, thereby supporting extinction memories.
Sigma-Aldrich
Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
BDNF human, Carrier free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
Anti-Brain Derived Neurotrophic Factor Antibody, Chemicon®, from sheep
Sigma-Aldrich
ProBDNF human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)