MilliporeSigma
  • Presynaptic Ca²⁺-permeable AMPA-receptors modulate paired-pulse depression in nociceptive sensory synapses.
Presynaptic Ca²⁺-permeable AMPA-receptors modulate paired-pulse depression in nociceptive sensory synapses.
Neuroscience letters (2014-12-03)
Mariia S Shypshyna, Nikolai S Veselovsky
ABSTRACT
The role of Ca(2+)-permeable AMPA-receptors (CP-AMPARs) in the induction of paired-pulse depression (PPD) at glutamatergic nociceptive sensory synapses was examined in co-culture of rat's dorsal root ganglion and dorsal horn neurons. CP-AMPARs make a considerable contribution to excitatory postsynaptic currents recorded in DH neurons following action potential generation in nociceptive DRG neurons. Activation of CP-AMPARs during fast synaptic transmission induces strong PPD (with a 200 ms inter-pulse interval). Blockage of CP-AMPARs with Naspm (100 μM) results in most cases in a significant reduction of the PPD magnitude, also reversing to paired-pulse facilitation in some cases. However, loading of postsynaptic DH neurons with BAPTA (10 mM) did not alter the effect of Naspm (100 μM) on the paired-pulse ratio. Our data provide evidence that presynaptic CP-AMPARs in nociceptive sensory synapses regulate the magnitude of PPD and that postsynaptic Ca(2+)-permeable glutamatergic receptors are not required for PPD induction.