MilliporeSigma
  • Fibronectin enhances spinal cord astrocyte proliferation by elevating P2Y1 receptor expression.
Fibronectin enhances spinal cord astrocyte proliferation by elevating P2Y1 receptor expression.
Journal of neuroscience research (2014-04-02)
Maosheng Xia, Yue Zhu
ABSTRACT
After spinal cord injury (SCI), the formation of glial scar is a complex process that is attributed primarily to astrocytic proliferation, but the mechanism of astrocytes proliferation is still unclear. Fibronectin is a large extracellular glycoprotein that helps organize the matrix protein, and its main membrane receptor is the α5 β1 integrin subunit. In this study, fibronectin stimulated spinal cord astrocytic proliferation from two directions: fibronectin increased astrocytic proliferation via α5 β1 integrin receptor, and fibronectin upregulated the expression of P2Y1 receptor, and adenosine triphosphate (ATP) could enhance the astrocytic proliferation and induce more release of arachidonic acid and prostaglandin E2 via P2Y1. The upregulation of P2Y1 by fibronectin required [Ca2+]i and the activation of integrin link kinase (ILK) and Akt. We found that [Ca2+]i stimulated by fibronectin was α5 β1 integrin receptor dependent and that the phosphorylation of Akt or extracellular signal-regulated protein kinase (ERK1/2) induced by fibronectin mediated the activation of cAMP response element-binding protein (CREB) and signal transducer and activator of transcription 3 (Stat3). Our research suggests that the release of fibronectin and ATP could stimulate the spinal cord astrocytic proliferation after SCI, and the expression of P2Y1 increased by fibronectin would provide more sites for ATP, which could aggravate the proliferation and inflammation of spinal cord astrocytes.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sulindac, ≥98.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human P2RY1
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis