MilliporeSigma
  • Accurate analysis of formose reaction products by LC-UV: an analytical challenge.
Accurate analysis of formose reaction products by LC-UV: an analytical challenge.
Journal of chromatographic science (2013-02-05)
Thomas Zweckmair, Stefan Böhmdorfer, Anna Bogolitsyna, Thomas Rosenau, Antje Potthast, Senad Novalin
ABSTRACT
This study presents a liquid chromatography-ultraviolet (LC-UV) method for the accurate analysis of formose reaction products with regard to sum parameters. Limitations encountered in gas chromatography (e.g., thermal stability of analytes) were overcome. First, alditols are O-benzoylated and subjected to reversed-phase chromatography. They are then detected by UV and by electrospray ionization-mass spectrometry (ESI-MS) for validation purposes. Compared with standard 2,4-dinitrophenylhydrazine labeling, the accuracy of results from LC-UV is dramatically improved due to enhanced chromatographic resolution and validation by ESI-MS. For the first time ever, LC-UV may be used to accurately quantify 2-hydroxymethylglycerol, a branched chain alditol, D,L-glyceraldehyde, D,L-galactose, which represents the group of D,L-iditol/galactose/glucose/mannose and D,L-erythrose in formose reaction products.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Supelco
Sodium formate, analytical standard
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O