MilliporeSigma
  • Transarterial chemoembolization vs. radiofrequency ablation for the treatment of single hepatocellular carcinoma 2 cm or smaller.
Transarterial chemoembolization vs. radiofrequency ablation for the treatment of single hepatocellular carcinoma 2 cm or smaller.
The American journal of gastroenterology (2014-06-18)
Jong Woo Kim, Jin Hyoung Kim, Kyu-Bo Sung, Heung-Kyu Ko, Ji Hoon Shin, Pyo Nyun Kim, Hyun-Kyung Choi, Gi-Young Ko, Hyun-Ki Yoon, Seng-Yong Chun, Dong Il Gwon
ABSTRACT
To compare the effectiveness of transarterial chemoembolization (TACE) and radiofrequency ablation (RFA) for treating small (≤2 cm) hepatocellular carcinomas (HCCs). This retrospective study consisted of 287 patients (mean age, 57.1 years; age range, 29-84 years; 221 men, 66 women; 73.5% with HBV; 100% with liver cirrhosis) with Barcelona Clinic Liver Cancer very early-stage HCC (≤2 cm single HCC) who were initially treated with TACE (n=122) or RFA (n=165). The primary study end point was overall patient survival. Secondary study end points were time to progression and tumor response. The RFA and TACE groups were well balanced in terms of baseline variables. The two groups did not differ significantly in overall survival (P=0.079) or major complication (P>0.999) rates. The respective cumulative survival rates at 1, 3, 5, and 8 years were 97.6, 86.7, 74.5, and 60.0% for RFA and 93.4, 75.4, 63.1, and 51.1% for TACE. Their objective tumor regression (complete or partial response) rates were 100% (165/165) and 95.9% (117/122), respectively (P=0.013). The median times to progression for RFA and TACE were 27.0±3.8 (95% confidence intervals (CIs): 19.6-34.4) and 18.0±2.9 (95% CIs: 12.2-23.8) months, respectively. RFA yielded a significantly longer time to progression (P=0.034). TACE may be a viable alternative treatment for ≤2 cm HCCs when RFA is not feasible.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard