MilliporeSigma
A universal approach to the synthesis of nanodendrites of noble metals.
Nanoscale (2014-05-06)
Yan Feng, Xiaohong Ma, Lin Han, Zhijian Peng, Jun Yang
ABSTRACT
Nanomaterials usually exhibit structure-dependent catalytic activity, selectivity, and stability. Herein, we report a universal approach for the synthesis of noble metal nanoparticles with a dendritic structure, which is based on the reduction of metal acetylacetonate precursors in oleylamine at a temperature of 160 °C. In this strategy, the metal acetylacetonate precursors are reduced into metal atoms by oleylamine and grow into metal nanoparticles, while oleylamine is simultaneously converted into oleylamide to protect the nanoparticles. The competition between particle aggregation and oleylamide passivation is essential to the formation of a large number of particle aggregates, which eventually grow into nanodendrites via Ostwald ripening process. In particular, in comparison with commercial PtRu/C catalysts, the alloy PtRuOs nanodendrites exhibited superior catalytic activity toward methanol oxidation.
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Ruthenium black
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
SAFC
Oleylamine, ≥98% (primary amine)
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Oleylamine, technical grade, 70%