MilliporeSigma
  • STAT3 represents a molecular switch possibly inducing astroglial instead of oligodendroglial differentiation of oligodendroglial progenitor cells in Theiler's murine encephalomyelitis.
STAT3 represents a molecular switch possibly inducing astroglial instead of oligodendroglial differentiation of oligodendroglial progenitor cells in Theiler's murine encephalomyelitis.
Neuropathology and applied neurobiology (2014-03-13)
Yanyong Sun, Annika Lehmbecker, Arno Kalkuhl, Ulrich Deschl, Wenhui Sun, Karl Rohn, Iva D Tzvetanova, Klaus-Armin Nave, Wolfgang Baumgärtner, Reiner Ulrich
ABSTRACT
Insufficient oligodendroglial differentiation of oligodendroglial progenitor cells (OPCs) is suggested to be responsible for remyelination failure and astroglial scar formation in Theiler's murine encephalomyelitis (TME). The aim of the present study is to identify molecular key regulators of OPC differentiation in TME, and to dissect their mechanism of action in vitro. TME virus (TMEV) infected SJL/J-mice were evaluated by rotarod analysis, histopathology, immunohistology and gene expression microarray analysis. The STAT3 pathway was activated using meteorin and inhibited using STAT3 inhibitor VII in the glial progenitor cell line BO-1 and in primary rat OPCs in vitro. As expected, immunohistology demonstrated progressively decreasing myelin basic protein-positive white matter in TME. In contrast, intralesional NG2-positive OPCs as well as GFAP-positive astrocytes were increased. Gene Set Enrichment Analysis revealed 26 Gene Ontology terms including 'JAK-STAT cascade' to be significantly positively correlated with the density of NG2-positive OPCs. Immunohistology revealed an increased amount of activated, phosphorylated STAT3-expressing astrocytes, OPCs, and microglia/macrophages within the lesions. Meteorin-induced activation of STAT3-signalling in BO-1 cells and primary rat OPCs resulted in an enhanced GFAP and reduced CNPase expression. In contrast, an oppositional result was observed in BO-1 cells treated with STAT3 inhibitor VII. The STAT3 pathway is a key regulator of OPC-differentiation, suggested to shift their differentiation from an oligodendroglial towards an astrocytic fate, thereby inducing astrogliosis and insufficient remyelination in TME.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Rat serum
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester, BioReagent, suitable for fluorescence, mixture of isomers, ≥80% (HPLC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O