MilliporeSigma
  • B cells regulate CD4+ T cell responses to papain following B cell receptor-independent papain uptake.
B cells regulate CD4+ T cell responses to papain following B cell receptor-independent papain uptake.
Journal of immunology (Baltimore, Md. : 1950) (2014-06-15)
Daniel F Dwyer, Matthew C Woodruff, Michael C Carroll, K Frank Austen, Michael F Gurish
ABSTRACT
Papain, a cysteine protease allergen with inherent adjuvant activity, induces potent IL-4 expression by T cells in the popliteal lymph nodes of mice following footpad immunization. In this study, we identify a novel, non-BCR-mediated capacity for B cells to rapidly bind and internalize papain. B cells subsequently regulate the adaptive immune response by enhancing ICOS expression on CD4(+) T cells and amplifying Th2 and follicular helper T cell induction. Ab blockade of ICOS ligand, expressed by popliteal lymph node B cells, but not dendritic cells, at the peak of the response inhibits IL-4 responses in wild-type mice but not B cell-deficient mice. Thus, B cells play a critical role in amplifying adjuvant-dependent Th2 polarization following noncanonical acquisition and internalization of the cysteine protease papain.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar