MilliporeSigma
  • Dynamic 3D analysis of myocardial sympathetic innervation: an experimental study using 123I-MIBG and a CZT camera.
Dynamic 3D analysis of myocardial sympathetic innervation: an experimental study using 123I-MIBG and a CZT camera.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2015-01-31)
Assuero Giorgetti, Silvia Burchielli, Vincenzo Positano, Gil Kovalski, Angela Quaranta, Dario Genovesi, Manuel Tredici, Valerio Duce, Luigi Landini, Maria Giovanna Trivella, Paolo Marzullo
ABSTRACT
Data on the in vivo myocardial kinetics of (123)I-metaiodobenzylguanidine ((123)I-MIBG) are scarce and have always been obtained using planar acquisitions. To clarify the normal kinetics of (123)I-MIBG in vivo over time, we designed an experimental protocol using a 3-dimensional (3D) dynamic approach with a cadmium zinc telluride (CZT) camera. We studied 6 anesthetized pigs (mean body weight, 37 ± 4 kg). Left ventricular myocardial perfusion and sympathetic innervation were assessed using (99m)Tc-tetrofosmin (26 ± 6 MBq), (123)I-MIBG (54 ± 14 MBq), and a CZT camera. A normal perfusion/function match on gated SPECT was the inclusion criterion. A dynamic acquisition in list mode started simultaneously with the bolus injection of (123)I-MIBG, and data were collected every 5 min for the first 20 min and then at acquisition steps of 30, 60, 90, and 120 min. Each step was reconstructed using dedicate software and reframed (60 s/frame). On the reconstructed transaxial slice that best showed the left ventricular cavity, regions of interest were drawn to obtain myocardial and blood pool activities. Myocardial time-activity curves were generated by interpolating data between contiguous acquisition steps, corrected for radiotracer decay and injected dose, and fitted to a bicompartmental model. Time to myocardial maximum signal intensity (MSI), MSI value, radiotracer retention index (RI, myocardial activity/blood pool integral), and washout rate were calculated. The mediastinal signal was measured and fitted to a linear model. The myocardial MSI of (123)I-MIBG was reached within 5.57 ± 4.23 min (range, 2-12 min). The mean MSI was 0.426% ± 0.092%. Myocardial RI decreased over time and reached point zero at 176 ± 31 min (range, 140-229 min). The ratio between myocardial and mediastinal signal at 15 and 125 min and extrapolated at 176 and 4 h was 5.45% ± 0.61%, 4.33% ± 1.23% (not statistically significant vs. 15 min), 3.95% ± 1.46% (P < 0.03 vs. 125 min), and 3.63% ± 1.64% (P < 0.03 vs. 176 min), respectively. Mean global washout rate at 125 min was 15% ± 14% (range, 0%-34%), and extrapolated data at 176 min and 4 h were 18% ± 18% (range, 0.49%-45%) and 25% ± 23% (range, 1.7%-56.2%; not statistically significant vs. 176 min), respectively. 3D dynamic analysis of (123)I-MIBG suggests that myocardial peak uptake is reached more quickly than previously described. Myocardial RI decreases over time and, on average, is null about 3 h after injection. The combination of an early peak and variations in delayed myocardial uptake could result in a wide physiologic range of washout rates. Mediastinal activity appears to be constant over time and significantly lower than previously described in planar studies, resulting in a higher heart-to-mediastinum ratio.
Product Description
Supelco
Azaperone, VETRANAL®, analytical standard
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Azaperone, European Pharmacopoeia (EP) Reference Standard
Atropine, European Pharmacopoeia (EP) Reference Standard