MilliporeSigma
  • Effect of telomerase inhibition on preclinical models of malignant rhabdoid tumor.
Effect of telomerase inhibition on preclinical models of malignant rhabdoid tumor.
Cancer genetics (2014-12-03)
Yafang Hu, Daniel Bobb, Yunbiao Lu, Jianping He, Jeffrey S Dome
ABSTRACT
Novel treatment approaches are desperately needed for malignant rhabdoid tumor (MRT). Telomerase is an attractive therapeutic target because it is specific to cancer and critical for cancer cell immortality. We evaluated the effect of the telomerase inhibitor imetelstat in preclinical models of MRT. Three MRT cell lines, BT-12, G401, and RT-peri, were treated with the telomerase inhibitor imetelstat. The effects of imetelstat on telomere length, DNA damage response, and cell proliferation were assessed. The efficacy of imetelstat in vivo was evaluated in subcutaneous xenografts derived from each of the cell lines. Treatment with imetelstat resulted in inhibition of telomerase activity, marked telomere shortening, and activation of the DNA damage response pathway, as measured by formation of γ-H2AX nuclear foci, phosphorylation of ATM, and phosphorylation of TP53. Imetelstat-treated G401 cells underwent complete growth arrest after 16 passages. The other two cell lines exhibited growth inhibition. Imetelstat resulted in 40-50% growth inhibition compared to placebo-treated controls in all three xenograft models. The activity of imetelstat as a single agent suggests that further studies of telomerase inhibitors in combination with other agents may be warranted.
Product Description
Supelco
Aphidicolin, analytical standard
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material