MilliporeSigma
  • CD45RA-Foxp3(low) non-regulatory T cells in the CCR7-CD45RA-CD27+CD28+ effector memory subset are increased in synovial fluid from patients with rheumatoid arthritis.
CD45RA-Foxp3(low) non-regulatory T cells in the CCR7-CD45RA-CD27+CD28+ effector memory subset are increased in synovial fluid from patients with rheumatoid arthritis.
Cellular immunology (2014-06-22)
Fumichika Matsuki, Jun Saegusa, Keisuke Nishimura, Yasushi Miura, Masahiro Kurosaka, Shunichi Kumagai, Akio Morinobu
ABSTRACT
Increased numbers of regulatory T (Treg) cells are found in synovial fluid from patients with rheumatoid arthritis (RASF) compared with peripheral blood. However, Treg cells in RASF have been shown to have a decreased capacity to suppress T cells. Here we phenotypically classified CD4+ T cells in RASF into six subsets based on the expression of CD45RA, CCR7, CD27 and CD28, and demonstrated that the CCR7-CD45RA-CD27+CD28+ TEM subset was significantly increased in synovial fluid compared with peripheral blood. In addition, the proportion of Foxp3+ Treg cells in the CCR7-CD45RA-CD27+CD28+ TEM subset was significantly increased in RASF. Furthermore, most of the Foxp3+ Treg cells in RASF were non-suppressive CD45RA-Foxp3(low) non-Treg cells, and the frequency of the non-Treg cells in the CCR7-CD45RA-CD27+CD28+ TEM subset was significantly increased in RASF. Our findings suggest that the pro-inflammatory environment in RA joints may induce the increase of CD45RA-Foxp3(low) non-Treg cells in synovial fluid.