MilliporeSigma
  • Characterisation of fatty acid, carotenoid, tocopherol/tocotrienol compositions and antioxidant activities in seeds of three Chenopodium quinoa Willd. genotypes.
Characterisation of fatty acid, carotenoid, tocopherol/tocotrienol compositions and antioxidant activities in seeds of three Chenopodium quinoa Willd. genotypes.
Food chemistry (2014-12-23)
Yao Tang, Xihong Li, Peter X Chen, Bing Zhang, Marta Hernandez, Hua Zhang, Massimo F Marcone, Ronghua Liu, Rong Tsao
ABSTRACT
Composition of fatty acids, tocopherols, tocotrienols, and carotenoids, and their contribution to antioxidant activities were investigated in seeds of three coloured quinoa cultivars (white, red and black). The major components and individual compounds were significantly different, and their concentrations were higher in darker seeds (p < 0.05). The oil yield was 6.58-7.17% which contained predominantly unsaturated fatty acids (89.42%). The ratio of omega-6/omega-3 fatty acid was ca. 6/1. The total tocopherol content ranged from 37.49 to 59.82 μg/g and mainly consisted of γ-tocopherol. Trace amount of α- and β-tocotrienols was also found. Black quinoa had the highest vitamin E followed by red and white quinoas. Carotenoids, mainly trans-lutein (84.7-85.6%) and zeaxanthin were confirmed for the first time in quinoa seeds, and the concentration was also the highest in black seeds. The antioxidant activities of lipophilic extracts were positively correlated with polyunsaturated fatty acids, total carotenoids and total tocopherols.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)
Fluorescein, European Pharmacopoeia (EP) Reference Standard