MilliporeSigma
  • Natural killer T cells are essential for the development of contact hypersensitivity in BALB/c mice.
Natural killer T cells are essential for the development of contact hypersensitivity in BALB/c mice.
The Journal of investigative dermatology (2014-04-24)
Chihiro Shimizuhira, Atsushi Otsuka, Tetsuya Honda, Akihiko Kitoh, Gyohei Egawa, Saeko Nakajima, Chisa Nakashima, Hiroshi Watarai, Yoshiki Miyachi, Kenji Kabashima
ABSTRACT
Contact hypersensitivity (CHS) has been widely used to study cutaneous immune responses, as a prototype of delayed-type hypersensitivity. Although natural killer T (NKT) cells have been assumed to have an important role in CHS, their role is controversial. Here, we report the role of NKT cells in the sensitization phase of CHS, by promoting the survival and maturation of dendritic cells (DCs) in the draining lymph nodes (LNs). The CHS response was attenuated with Cd1d1(-/-) and Traj18(-/-) BALB/c mice in which NKT cells were absent. In the draining LNs, the number of effector T cells and cytokine production were significantly reduced with NKT cell-deficient mice. NKT cells activated and colocalized with DCs in the draining LNs after sensitization. The number of migrated and mature DCs was reduced in NKT cell-deficient mice 72 hours after FITC application. In in vitro experiments, activated NKT cells enhanced bone marrow-derived DC (BMDC) survivability via tumor necrosis factor (TNF) production from BMDCs. In addition, TNF production from BMDCs was partially suppressed by the neutralizing anti-CD54 or CD154 antibodies. Our data demonstrate that DC-NKT interaction has a pivotal role in the sensitization phase of CHS.
Sigma-Aldrich
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dibutyl phthalate, 99%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Mouse IL-13 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-13 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Rat IL-13 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
DBP, 98% (HPLC)
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, purum p.a., ≥98.0% (GC)
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)