MilliporeSigma
  • Enantioselective high performance liquid chromatography and supercritical fluid chromatography separation of spirocyclic terpenoid flavor compounds.
Enantioselective high performance liquid chromatography and supercritical fluid chromatography separation of spirocyclic terpenoid flavor compounds.
Journal of chromatography. A (2014-07-22)
Mathias Schaffrath, Verena Weidmann, Wolfgang Maison
ABSTRACT
Chiral spirocyclic terpenoids are abundant natural flavors with significant impact particularly on the food industry. Chromatographic methods for analytical and preparative separation of these compounds are therefore of high interest to natural product chemists in academia and industry. Gas chromatography on chiral stationary phases is currently the standard method for the separation of volatile terpenoids, limiting the scale to analytical quantities. We report herein high performance liquid chromatography (HPLC) and supercritical fluid chromatography (SFC) protocols for the chiral separation of several racemic spirocyclic terpenoids such as the important flavors theaspirane and vitispirane. A screening of mobile phases and 16 commercially available chiral stationary phases (CSPs) largely based on polysaccharides led to identification of protocols for the separation of all terpenoids tested. SFC methods were found to be particularly useful for the separation of these spirocyclic flavors due to the volatility and low polarity of the compounds. The reported chiral HPLC and SFC protocols are scalable alternatives to gas chromatographic separations of volatile terpenoid flavors.
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Heptane, anhydrous, 99%
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%