MilliporeSigma
  • High neuropeptide Y release associates with Ewing sarcoma bone dissemination - in vivo model of site-specific metastases.
High neuropeptide Y release associates with Ewing sarcoma bone dissemination - in vivo model of site-specific metastases.
Oncotarget (2015-02-26)
Sung-Hyeok Hong, Jason U Tilan, Susana Galli, Ewa Izycka-Swieszewska, Taylor Polk, Meredith Horton, Akanksha Mahajan, David Christian, Shari Jenkins, Rachel Acree, Katherine Connors, Phuong Ledo, Congyi Lu, Yi-Chien Lee, Olga Rodriguez, Jeffrey A Toretsky, Chris Albanese, Joanna Kitlinska
ABSTRACT
Ewing sarcoma (ES) develops in bones or soft tissues of children and adolescents. The presence of bone metastases is one of the most adverse prognostic factors, yet the mechanisms governing their formation remain unclear. As a transcriptional target of EWS-FLI1, the fusion protein driving ES transformation, neuropeptide Y (NPY) is highly expressed and released from ES tumors. Hypoxia up-regulates NPY and activates its pro-metastatic functions. To test the impact of NPY on ES metastatic pattern, ES cell lines, SK-ES1 and TC71, with high and low peptide release, respectively, were used in an orthotopic xenograft model. ES cells were injected into gastrocnemius muscles of SCID/beige mice, the primary tumors excised, and mice monitored for the presence of metastases. SK-ES1 xenografts resulted in thoracic extra-osseous metastases (67%) and dissemination to bone (50%) and brain (25%), while TC71 tumors metastasized to the lungs (70%). Bone dissemination in SK-ES1 xenografts associated with increased NPY expression in bone metastases and its accumulation in bone invasion areas. The genetic silencing of NPY in SK-ES1 cells reduced bone degradation. Our study supports the role for NPY in ES bone invasion and provides new models for identifying pathways driving ES metastases to specific niches and testing anti-metastatic therapeutics.
Sigma-Aldrich
Nitrous oxide, 99%
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human NPY
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Npy