MilliporeSigma
  • The upregulation of LAG-3 on T cells defines a subpopulation with functional exhaustion and correlates with disease progression in HIV-infected subjects.
The upregulation of LAG-3 on T cells defines a subpopulation with functional exhaustion and correlates with disease progression in HIV-infected subjects.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-18)
Xiaoling Tian, Anli Zhang, Chao Qiu, Wei Wang, Yu Yang, Chenli Qiu, Aiping Liu, Lingyan Zhu, Songhua Yuan, Huiliang Hu, Wanhai Wang, Qiang Wei, Xiaoyan Zhang, Jianqing Xu
ABSTRACT
T cells develop functional defects during HIV-1 infection, partially due to the upregulation of inhibitory receptors such as programmed death-1 (PD-1) and CTLA-4. However, the role of lymphocyte activation gene-3 (LAG-3; CD223), also known as an inhibitory receptor, in HIV infection remains to be determined. In this study, we revealed that LAG-3 on T cells delivers an inhibitory signal to downregulate T cell functionality, thereby playing an immunoregulatory role during persistent HIV-1 infection. We observed that HIV-1 infection results in a significant increase in LAG-3 expression in both the peripheral blood and the lymph nodes. The upregulation of LAG-3 is dramatically manifested on both CD4(+) and CD8(+) T cells and is correlated with disease progression. As expected, prolonged antiretroviral therapy reduces the expression of LAG-3 on both CD4(+) and CD8(+) T cells. The ex vivo blockade of LAG-3 significantly augments HIV-specific CD4(+) and CD8(+) T cell responses, whereas the overexpression of LAG-3 in T cells or the stimulation of LAG-3 on T cells leads to the reduction of T cell responses. Furthermore, most LAG-3 and PD-1 are expressed in different T cell subsets. Taken together, these data demonstrate that the LAG-3/MHC class II pathway plays an immunoregulatory role, thereby providing an important target for enhancing immune reconstitution in HIV-infected patients. Additionally, the LAG-3/MHC class II pathway may synergize with PD-1/PD ligand to enhance T cell-mediated immune responses.
Sigma-Aldrich
LAG-3 (human): FC (human), recombinant, expressed in CHO cells, >99% (SDS-PAGE)
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture