MilliporeSigma
  • Effects of altering alfalfa hay quality when feeding steam-flaked versus high-moisture corn grain on ruminal fermentation and lactational performance of dairy cows.
Effects of altering alfalfa hay quality when feeding steam-flaked versus high-moisture corn grain on ruminal fermentation and lactational performance of dairy cows.
Journal of dairy science (2014-09-30)
J-S Eun, A W Kelley, K Neal, A J Young, J O Hall
ABSTRACT
This experiment was performed to test a hypothesis that nutritive benefits of feeding high-moisture corn (HMC) would be different when fed with different qualities of alfalfa hay (AH) due to associative effects on ruminal fermentation and nutrient utilization efficiency. Eight multiparous lactating Holstein cows were used; 4 were surgically fitted with ruminal cannulas. Days in milk averaged 184 ± 10.7 at the start of the experiment. The experiment was performed in a duplicate 4 × 4 Latin square design. Within each square, cows were randomly assigned to a sequence of 4 diets during each of the four 21-d periods (14 d of treatment adaptation and 7 d of data collection and sampling). A 2 × 2 factorial arrangement was used; fair-quality AH [FAH; 39.6% neutral detergent fiber (NDF) and 17.9% crude protein (CP)] or high-quality AH (HAH; 33.6% NDF and 21.9% CP) was combined with steam-flaked corn (SFC) or HMC to form 4 treatments: FAH with SFC, FAH with HMC, HAH with SFC, and HAH with HMC. The AH was fed at 32% dry matter (DM) content, whereas SFC or HMC was included at 17% DM content. Quality of AH did not affect DM intake, whereas feeding HMC decreased DM intake, regardless of quality of AH. Digestibility of DM was greater for cows fed HAH compared with those fed FAH (70.1 vs. 67.6%). Digestibility of NDF increased by feeding HMC (67.6 vs. 58.4%), but not by quality of AH. Under FAH, starch digestibility decreased by feeding HMC compared with SFC (85.7 vs. 95.0%), but it was similar under HAH, resulting in an interaction between quality of AH and type of corn grain (CG). Feeding different qualities of AH did not affect milk yield; however, feeding HMC decreased milk yield in FAH diet, causing an AH × CG interaction. Efficiency of milk yield/DM intake was improved due to feeding HMC, regardless of the quality of the AH. In addition, dietary N utilization for milk N tended to increase by feeding HMC, but it was not influenced by quality of AH. Yield of microbial protein increased by feeding HAH diets compared with FAH diets, whereas feeding the HMC diet increased microbial protein yield under the HAH diet, leading to an interaction between and AH and CG. Overall results in this experiment indicate that feeding HMC in AH-based diets improved feed efficiency as well as N utilization efficiency, regardless of quality of AH.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Sigma-Aldrich
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Sodium sulfite, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium sulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
Sigma-Aldrich
Sodium sulfite, puriss. p.a., reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Sigma-Aldrich
Sodium sulfite, ≥98%
Supelco
Sodium sulfite, analytical standard