MilliporeSigma
  • Effect of dutasteride in men receiving intermittent androgen ablation therapy: The AVIAS trial.
Effect of dutasteride in men receiving intermittent androgen ablation therapy: The AVIAS trial.
Canadian Urological Association journal = Journal de l'Association des urologues du Canada (2014-12-09)
Laurence Klotz, Abdenour Nabid, Celestia Higano, Chris Ryanm, Marlene Kebabdjian, Joseph Chin
ABSTRACT
We studied the effect of dutasteride on the length of the off-treatment period in prostate cancer patients on intermittent androgen deprivation (IAD) therapy. We conducted a randomized, placebo-controlled Phase II trial in men with localized prostate cancer and a rising prostate-specific antigen (PSA) level post-primary treatment. Patients were randomized to dutasteride (0.5 mg/day) or placebo. All patients received androgen deprivation therapy (ADT), which was stopped at month 9 if the PSA level was <1.0 ng/mL. ADT was resumed when PSA increased to ≥5.0 ng/mL. End points included time off treatment, PSA nadir after 9 months of ADT, serum testosterone and dihydrotestosterone levels, and time to castrate-resistant prostate cancer (rising PSA while testosterone levels remain <50 ng/mL). There were 87 evaluable patients: 49 dutasteride, 38 placebo. In total, 80 patients completed one treatment cycle: 45 dutasteride, 35 placebo. The median time off treatment for patients reaching ≥5 ng/mL was 18.6 and 16.7 months for dutasteride and placebo, respectively (p = 0.7600). The median PSA nadir at 9 months was 0.1 and 0.075 ng/mL, respectively (p = 0.4486). There were no cases of androgen-independent prostate cancer. Our study limitations include its short duration with only one treatment cycle evaluated. This small-scale Phase II randomized controlled trial showed no benefit to the addition of dutasteride to an IAD regimen.
Product Description
Dutasteride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.