MilliporeSigma
  • Metabolite profiling of anhuienoside C by rat intestinal bacteria using the LC-MS metabolomic approach.
Metabolite profiling of anhuienoside C by rat intestinal bacteria using the LC-MS metabolomic approach.
Xenobiotica; the fate of foreign compounds in biological systems (2014-09-16)
Huinan Zhao, Hui Liu, Zhiguo Ma, Ying Wang, Yao-Lan Li, Wen-Cai Ye, Baojian Wu
ABSTRACT
1.Anhuienoside C (A-C) is the main active component of the saponin exact of "Di Wu", an oral drug for rheumatism treatment in China. In this study, we aimed to elucidate the metabolic pathways of A-C by intestinal bacteria using the metabolomic approach. 2.Four deglycosylated metabolites (M1, M2, M3 and M4) were identified after A-C (50 µM) was incubated with rat fecal lysate. Chemical structures of these metabolites were determined by high-resolution masses and nuclear magnetic resonance (NMR). 3.A one-compartment pharmacokinetic model was used to describe the formation of bacterial metabolites at a dose of 10 µM A-C. The results revealed that formation of M1 and M2 was rapid, whereas formation of M3 was rather slow. Further, it was found that the metabolites were generated by successive cleavage of the glycosyl residues. 4.This is the first report that A-C is subjected to efficient bacterial metabolism in the gut with M1 and M2 as main metabolites. Our study should be helpful for a better understanding of in vivo disposition of oral A-C.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Oleanolic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Oleanolic acid, ≥97%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard