MilliporeSigma
  • A novel function for lysyl oxidase in pluripotent mesenchymal cell proliferation and relevance to inflammation-associated osteopenia.
A novel function for lysyl oxidase in pluripotent mesenchymal cell proliferation and relevance to inflammation-associated osteopenia.
PloS one (2014-06-28)
Roozbeh Khosravi, Katharine L Sodek, Wan-Peng Xu, Manish V Bais, Debashree Saxena, Michael Faibish, Philip C Trackman
ABSTRACT
Lysyl oxidase is a multifunctional enzyme required for collagen biosynthesis. Various growth factors regulate lysyl oxidase during osteoblast differentiation, subject to modulation by cytokines such as TNF-α in inflammatory osteopenic disorders including diabetic bone disease. Canonical Wnt signaling promotes osteoblast development. Here we investigated the effect of Wnt3a and TNF-α on lysyl oxidase expression in pluripotent C3H10T1/2 cells, bone marrow stromal cells, and committed osteoblasts. Lysyl oxidase was up-regulated by a transcriptional mechanism 3-fold in C3H10T1/2 cells, and 2.5-fold in bone marrow stromal cells. A putative functional TCF/LEF element was identified in the lysyl oxidase promoter. Interestingly, lysyl oxidase was not up-regulated in committed primary rat calvarial- or MC3T3-E1 osteoblasts. TNF-α down-regulated lysyl oxidase both in Wnt3a-treated and in non-treated C3H10T1/2 cells by a post-transcriptional mechanism mediated by miR203. Non-differentiated cells do not produce a collagen matrix; thus, a novel biological role for lysyl oxidase in pluripotent cells was investigated. Lysyl oxidase shRNAs effectively silenced lysyl oxidase expression, and suppressed the growth of C3H10T1/2 cells by 50%, and blocked osteoblast differentiation. We propose that interference with lysyl oxidase expression under excess inflammatory conditions such as those that occur in diabetes, osteoporosis, or rheumatoid arthritis can result in a diminished pool of pluripotent cells which ultimately contributes to osteopenia.
Sigma-Aldrich
MISSION® esiRNA, targeting human LOX