MilliporeSigma
  • Interleukin-10 blocks in vitro replication of human cytomegalovirus by inhibiting the virus-induced autophagy in MRC5 cells.
Interleukin-10 blocks in vitro replication of human cytomegalovirus by inhibiting the virus-induced autophagy in MRC5 cells.
Biochemical and biophysical research communications (2014-05-07)
Li Wang, Huiping Zhang, Jihong Qian, Kanqing Wang, Jianxing Zhu
ABSTRACT
Interleukin-10 is an important cytokine that regulates immune response. Previous studies have shown that human cytomegalovirus can trigger cell autophagy during the early stages of infection. To our knowledge, whether IL-10 inhibits HCMV-induced autophagy and virus replication has not been studied previously. We investigated whether IL-10 affects cell viability and autophagy under the conditions of starvation and HCMV infection by using the MRC5 cell line. We also explored the role of IL-10-mediated autophagy on HCMV replication. Our data showed that IL-10 inhibited the autophagic flux of the MRC5 cells irrespective of starvation or HCMV infection, and suppressed HCMV replication. The promotion of autophagy with either a pharmacological inducer (rapamycin), or a technique to over-express the BECN1 gene reversed the effect of IL-10 on virus replication. Furthermore, the PI3K/Akt signal pathway was activated when the cells were pretreated with IL-10. Our results indicated that IL-10 can suppress HCMV replication by inhibiting autophagy in host cells during the early stages of infection.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining