MilliporeSigma
First fully automated immunoassay for anti-Müllerian hormone.
Clinical chemistry and laboratory medicine (2014-03-14)
Dieter Gassner, Rebecca Jung
ABSTRACT
The increasing importance of anti-Müllerian hormone (AMH) for the assessment of ovarian reserve requires accurate AMH measurements. There have been conflicting results about the reliability of currently existing manual AMH assays. Development of a high sensitive, fast and fully automated AMH assay on the Elecsys®/cobas e electrochemiluminescence immunoassay platform. Elecsys® AMH is a monoclonal two-site assay used to measure AMH in 50 µL of serum or lithium heparin plasma in about 18 min. Its measuring range is from 0.01 to 23 ng/mL. The assay detects primarily 140 kDa total AMH (proAMH and AMHN,C). Standardization was against the Beckman AMH Gen II. Recovery against the highly cited Immunotech calibration was about 90%. Within-run imprecision coefficient of variation (CV) calculated on 10 serum samples were between 0.5% and 1.8%. Repeatability and intermediate precision calculated on 14 serum samples ranged from 2.6% to 1.7% and 2.9% to 2.1%, respectively. Limit of detection (LoD) [limit of quantitation (LoQ)] was 0.01 ng/mL (0.03 ng/mL). Percent recovery in dilution studies was <10% and after mixing of high and low AMH pools 94% to 103%. Elecsys® AMH, when compared to revised AMH Gen II (or Ansh Labs ultrasensitive AMH ELISA) using 57 female serum samples yielded a correlation coefficient of 0.98 (0.97) and a slope of 0.81 (0.73). There was no evidence for sample instability or variability. Samples stored at 20-25 °C or 2-8 °C up to 7 days showed no significant storage issues. Freeze-thaw cycles or sample storage at -20 °C and -80 °C for up to 9 months was without any effect on measured AMH. Availability of an automated Elecsys® AMH assay offers an attractive alternative to the current manual AMH assays.
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%