MilliporeSigma
  • Cytokine profile in cervical mucosa of Japanese patients with cervical intraepithelial neoplasia.
Cytokine profile in cervical mucosa of Japanese patients with cervical intraepithelial neoplasia.
International journal of clinical oncology (2014-03-01)
Takashi Iwata, Takuma Fujii, Kenji Morii, Miyuki Saito, Juri Sugiyama, Hiroshi Nishio, Tohru Morisada, Kyoko Tanaka, Tomonori Yaguchi, Yutaka Kawakami, Daisuke Aoki
ABSTRACT
Immune responses in the uterine cervix are considered to play an important role in persistent human papillomavirus (HPV) infection and carcinogenesis, but many aspects of the mechanism are still unclear. The goal of this study was to measure cytokines to analyze immune responses in patients with cervical intraepithelial neoplasia (CIN). The levels of 17 cytokines (IL-1β, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12, IL-13, IL-17, G-CSF, GM-CSF, INF-γ, MCP-1, MIP-1β, and TNFα) in cervical mucus were simultaneously measured using a multiplex immunoassay in 52 high-grade squamous intraepithelial lesion (HSIL) cases and overproduction of IL-1β, IL-8, and MIP-1β was identified. The levels of these 3 cytokines were measured in 130 patients with or without CIN lesions using enzyme-linked immunosorbent assay. The associations of the cytokine levels with the cytology, infecting HPV type, and status of cigarette smoking were investigated. IL-1β and IL-8 levels were associated with the cytology, and these levels were higher in HSIL cases than in NILM (negative for intraepithelial lesion and malignancy) and LSIL (low-grade squamous intraepithelial lesion) cases (P = 0.005, P = 0.001, respectively). The MIP-1β level was significantly lower in smokers (P = 0.018) and high-risk (HR)-HPV-infected patients (P = 0.021). Enhanced expression of IL-1β and IL-8 indicates that Th2 inflammatory responses become stronger in the local uterine cervical region with the progression of CIN lesions, and a decrease in the MIP-1β level may be advantageous for immunoescape of HPV. Cigarette smoking may further facilitate persistent HPV infection.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material