MilliporeSigma
  • De-differentiation confers multidrug resistance via noncanonical PERK-Nrf2 signaling.
De-differentiation confers multidrug resistance via noncanonical PERK-Nrf2 signaling.
PLoS biology (2014-09-10)
Catherine A Del Vecchio, Yuxiong Feng, Ethan S Sokol, Erik J Tillman, Sandhya Sanduja, Ferenc Reinhardt, Piyush B Gupta
ABSTRACT
Malignant carcinomas that recur following therapy are typically de-differentiated and multidrug resistant (MDR). De-differentiated cancer cells acquire MDR by up-regulating reactive oxygen species (ROS)-scavenging enzymes and drug efflux pumps, but how these genes are up-regulated in response to de-differentiation is not known. Here, we examine this question by using global transcriptional profiling to identify ROS-induced genes that are already up-regulated in de-differentiated cells, even in the absence of oxidative damage. Using this approach, we found that the Nrf2 transcription factor, which is the master regulator of cellular responses to oxidative stress, is preactivated in de-differentiated cells. In de-differentiated cells, Nrf2 is not activated by oxidation but rather through a noncanonical mechanism involving its phosphorylation by the ER membrane kinase PERK. In contrast, differentiated cells require oxidative damage to activate Nrf2. Constitutive PERK-Nrf2 signaling protects de-differentiated cells from chemotherapy by reducing ROS levels and increasing drug efflux. These findings are validated in therapy-resistant basal breast cancer cell lines and animal models, where inhibition of the PERK-Nrf2 signaling axis reversed the MDR of de-differentiated cancer cells. Additionally, analysis of patient tumor datasets showed that a PERK pathway signature correlates strongly with chemotherapy resistance, tumor grade, and overall survival. Collectively, these results indicate that de-differentiated cells up-regulate MDR genes via PERK-Nrf2 signaling and suggest that targeting this pathway could sensitize drug-resistant cells to chemotherapy.
Product Description
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Menadione, European Pharmacopoeia (EP) Reference Standard
USP
Menadione, United States Pharmacopeia (USP) Reference Standard
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material