MilliporeSigma
  • A simultaneous extraction method for organophosphate, pyrethroid, and neonicotinoid insecticides in aqueous samples.
A simultaneous extraction method for organophosphate, pyrethroid, and neonicotinoid insecticides in aqueous samples.
Archives of environmental contamination and toxicology (2015-01-23)
Chloé de Perre, Sara A Whiting, Michael J Lydy
ABSTRACT
A method was developed for the extraction and analysis of 2 organophosphate, 8 pyrethroid, and 5 neonicotinoid insecticides from the same water sample. A salted liquid-liquid extraction (LLE) was optimized with a solid-phase extraction (SPE) step that separated the organophosphates (OPs) and pyrethroids from the neonicotinoids. Factors that were optimized included volume of solvent and amount of salt used in the LLE, homogenization time for the LLE, and type and volume of eluting solvent used for the SPE. The OPs and pyrethroids were quantified using gas chromatography-mass spectrometry, and the neonicotinoids were quantified using liquid chromatography-diode array detector. Results showed that the optimized method was accurate, precise, reproducible, and robust; recoveries in river water spiked with 100 ng L(-1) of each of the insecticides were all between 86 and 114 % with RSDs between 2 and 8 %. The method was also sensitive with method detection limits ranging from 0.1 to 27.2 ng L(-1) depending on compounds and matrices. The optimized method was thus appropriate for the simultaneous extraction of 15 widely applied insecticides from three different classes and was shown to provide valuable information on their environmental fate from field-collected aqueous samples.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Acetic acid, analytical standard
Supelco
Acetone, analytical standard
Supelco
Methanol, analytical standard
Supelco
β-Cyfluthrin, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)