MilliporeSigma
  • Fast carbohydrate analysis via liquid chromatography coupled with ultra violet and electrospray ionization ion trap detection in 96-well format.
Fast carbohydrate analysis via liquid chromatography coupled with ultra violet and electrospray ionization ion trap detection in 96-well format.
Journal of chromatography. A (2014-05-28)
Broder Rühmann, Jochen Schmid, Volker Sieber
ABSTRACT
A fast carbohydrate screening platform processible in 96-well format is described. The method is suitable for the determination of various carbohydrates out of complex mixtures as obtained by acidic hydrolysis of carbohydrates polymers. The chromatographic conditions for an efficient separation (12min) and the derivatization process with 1-phenyl-3-methyl-5-pyrazolone (PMP) were optimized for high resolution separation and simultaneous determination of deoxy-, amino-, anhydro-sugars as well as hexoses, pentoses, dimers, uronic acids and degradation products like furfural and hydroxymethylfurfural (HMF). The potential to quantify with UV- and MS-detector in the same range has been demonstrated for 20 different compounds. Finally, the matrix effects of the hydrolysis were positively evaluated. The micro scale hydrolysis and PMP-derivatization without any extraction or drying steps, both in 96-well format, result in a fast and intuitive sample preparation. In combination with a fast liquid chromatography coupled to UV and electrospray ionization ion trap detection (LC-UV-ESI-MS/MS) for the qualification and quantification of various sugars, dimers and degradation products, this method shows great performance in carbohydrate analysis.
Product Description
Millipore
D-(+)-Cellobiose, ≥99.0%, suitable for microbiology, Reducing sugar disaccharide
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
D-Allose, 98%
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Ribose, suitable for cell culture, BioReagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Cellobiose, ≥98% (HPLC)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture