MilliporeSigma
  • Degrading a mixture of three textile dyes using photo-assisted electrochemical process with BDD anode and O₂-diffusion cathode.
Degrading a mixture of three textile dyes using photo-assisted electrochemical process with BDD anode and O₂-diffusion cathode.
Environmental science and pollution research international (2014-04-12)
Alireza Khataee, Mahdie Safarpour, Behrouz Vahid, Amaneh Akbarpour
ABSTRACT
In this paper, degradation of a mixture of three azo dyes was studied by the photo-assisted electrochemical process using an O₂-diffusion cathode containing carbon nanotubes and boron-doped diamond (BDD) anode. The concentration of three textile dyes (C.I. Acid Orange 8 (AO8), C.I. Acid Orange 10 (AO10), and C.I. Acid Orange 12 (AO12)) was determined simultaneously despite the severe overlap of their spectra. For this purpose, partial least square (PLS), as a multivariate calibration method, was utilized based on recording UV-Vis spectra during the decolorization process. Moreover, the central composite design was used for the modeling of photo-assisted electrochemical decolorization of the aqueous solutions containing three dyes. The investigated parameters were the initial concentration of three dyes, applied current and reaction time. Analysis of variance (ANOVA) revealed that the obtained regression models match the experimental results well with R (Khataee et al. 2010, Clean-Soil Air Water 38 (1):96-103, 2010) of 0.972, 0.971, and 0.957 for AO8, AO10, and AO12, respectively. Three-dimensional surface and contour plots were applied to describe the relation between experimental conditions and the observed response. The results of TOC analysis confirmed good ability of proposed photo-assisted electrochemical process for degradation and mineralization of textile industry wastewater.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%