MilliporeSigma
  • Low circulating 25-hydroxyvitamin D concentrations are associated with defects in insulin action and insulin secretion in persons with prediabetes.
Low circulating 25-hydroxyvitamin D concentrations are associated with defects in insulin action and insulin secretion in persons with prediabetes.
The Journal of nutrition (2015-03-06)
Fahim Abbasi, Christine Blasey, David Feldman, Michael P Caulfield, Feras M Hantash, Gerald M Reaven
ABSTRACT
Individuals with prediabetes mellitus (PreDM) and low circulating 25-hydroxyvitamin D [25(OH)D] are at increased risk of type 2 diabetes mellitus (T2DM). We aimed to determine whether low 25(OH)D concentrations are associated with defects in insulin action and insulin secretion in persons with PreDM. In this cross-sectional study, we stratified 488 nondiabetic subjects as having PreDM or normal fasting glucose (NFG) and a 25(OH)D concentration ≤20 ng/mL (deficient) or >20 ng/mL (sufficient). We determined insulin resistance by steady state plasma glucose (SSPG) concentration and homeostasis model assessment of insulin resistance (HOMA-IR) and insulin secretion by homeostasis model assessment of β-cell function (HOMA-β). We compared insulin resistance and secretion measures in PreDM and NFG groups; 25(OH)D-deficient and 25(OH)D-sufficient groups; and PreDM-deficient, PreDM-sufficient, NFG-deficient, and NFG-sufficient subgroups, adjusting for age, sex, race, body mass index, multivitamin use, and season. In the PreDM group, mean SSPG concentration and HOMA-IR were higher and mean HOMA-β was lower than in the NFG group (P < 0.001 for all comparisons). In the 25(OH)D-deficient group, mean SSPG concentration was higher (P < 0.001), but neither mean HOMA-IR nor HOMA-β was significantly different from that in the 25(OH)D-sufficient group. In the PreDM-deficient subgroup, mean (95% CI) SSPG concentration was higher (P < 0.01) than in the PreDM-sufficient, NFG-deficient, and NFG-sufficient subgroups [192 (177-207) mg/dL vs. 166 (155-177) mg/dL, 148 (138-159) mg/dL, and 136 (127-144) mg/dL, respectively]. Despite greater insulin resistance, mean HOMA-β was not significantly higher in the PreDM-deficient subgroup than in the PreDM-sufficient, NFG-deficient, and NFG-sufficient subgroups [98 (85-112) vs. 91 (82-101), 123 (112-136), and 115 (106-124), respectively]. Subjects with PreDM and low circulating 25(OH)D concentrations are the subgroup of nondiabetic individuals who are the most insulin resistant and have impaired β-cell function, attributes that put them at enhanced risk of T2DM.
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 50 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D2, ≥95.0% (HPLC)
Supelco
25-Hydroxyvitamin D2 solution, 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
25-Hydroxyvitamin D2 solution, 100 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D2 solution, 5 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D2 solution, 50 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Supelco
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Calcifediol, European Pharmacopoeia (EP) Reference Standard