MilliporeSigma
  • Seminal fluid promotes in vitro sperm-oviduct binding in the domestic cat (Felis catus).
Seminal fluid promotes in vitro sperm-oviduct binding in the domestic cat (Felis catus).
Theriogenology (2015-03-05)
F Henry, S Eder, K Reynaud, J Schön, G Wibbelt, A Fontbonne, K Müller
ABSTRACT
From many endangered or threatened species which are expected to profit from assisted reproduction techniques, mainly epididymal sperm of dead or freshly castrated males are available. These sperm had contact to epididymal secretion products but not to seminal fluid components. Notably, products of accessory sex glands have been shown in domestic animals to condition sperm for fertilization, in particular by mediating sperm-oviduct interaction. We report for the first time that motile epididymal sperm from domestic cats are able to bind to fresh oviduct epithelial cell explants from preovulatory females (median [min, max] of 10 [8, 16] and 10 [8, 17] sperm per 0.01 mm(2) explant surface from both isthmic and ampullar regions, respectively). More sperm attach to the explants when epididymal sperm were preincubated for 30 minutes with seminal fluid separated from electroejaculates of mature tomcats (median [min, max] of 17 [13, 25] and 16 [12, 21] sperm per 0.01 mm(2) explant surface from isthmus and ampulla, respectively). The proportion of bound sperm increased from a median of 54% to 62% by seminal fluid treatment. Sperm-oviduct binding could be facilitated by the decelerated sperm motion which was observed in seminal fluid-treated samples or supported by seminal fluid proteins newly attached to the sperm surface. Seminal fluid had no effect on the proportion of sperm with active mitochondria. Extent and pattern of sperm interaction in vitro were independent of explant origin from isthmus or ampulla. Sperm were attached to both cilia and microvilli of the main epithelial cell types present in all explants. In contrast to published sperm-binding studies with porcine and bovine oviduct explants where predominantly the anterior head region of sperm was attached to ciliated cells, the tails of some cat sperm were firmly stuck to the oviduct cell surfaces, whereas the heads were wobbling. Whether this response is a preliminary step toward phagocytosis or a precondition to capacitation and fertilization remains to be determined. In conclusion, treatment of epididymal sperm with seminal fluid or particular protein components should be considered in future investigations for its potential to improve the outcome of artificial insemination in felids.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Medium 199, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%