Merck
  • Untargeted profiling of urinary steroid metabolites after testosterone ingestion: opening new perspectives for antidoping testing.
Untargeted profiling of urinary steroid metabolites after testosterone ingestion: opening new perspectives for antidoping testing.
Bioanalysis (2014-11-21)
Julien Boccard, Flavia Badoud, Nicolas Jan, Raul Nicoli, Carine Schweizer, François Pralong, Jean-Luc Veuthey, Norbert Baume, Serge Rudaz, Martial Saugy
ABSTRACT
Antidoping procedures are expected to greatly benefit from untargeted metabolomic approaches through the discovery of new biomarkers of prohibited substances abuse. Endogenous steroid metabolites were monitored in urine samples from a controlled elimination study of testosterone undecanoate after ingestion. A platform coupling ultra-high pressure LC with high-resolution quadrupole TOF MS was used and high between-subject metabolic variability was successfully handled using a multiblock data analysis strategy. Links between specific subsets of metabolites and influential genetic polymorphisms of the UGT2B17 enzyme were highlighted. This exploratory metabolomic strategy constitutes a first step toward a better understanding of the underlying patterns driving the high interindividual variability of steroid metabolism. Promising biomarkers were selected for further targeted study.
Product Description
Supelco
Methanol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Absolute - Acetone free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O