MilliporeSigma
  • A case-control study evaluating the relationship between thimerosal-containing haemophilus influenzae type b vaccine administration and the risk for a pervasive developmental disorder diagnosis in the United States.
A case-control study evaluating the relationship between thimerosal-containing haemophilus influenzae type b vaccine administration and the risk for a pervasive developmental disorder diagnosis in the United States.
Biological trace element research (2014-11-11)
David A Geier, Janet K Kern, Paul G King, Lisa K Sykes, Mark R Geier
ABSTRACT
Thimerosal is an organic mercury (Hg)-containing compound (49.55 % Hg by weight) historically added to many multi-dose vials of vaccine as a preservative. A hypothesis testing case-control study evaluated automated medical records in the Vaccine Safety Datalink (VSD) for organic Hg exposure from Thimerosal in Haemophilus influenzae type b (Hib)-containing vaccines administered at specific times within the first 15 months of life among subjects diagnosed with pervasive developmental disorder (PDD) (n = 534) in comparison to controls. The generally accepted biologically non-plausible linkage between Thimerosal exposure and subsequent diagnosis of febrile seizure (n = 5886) was examined as a control outcome. Cases diagnosed with PDD received significantly more organic Hg within the first 6 months of life (odds ratio (OR) = 1.97, p < 0.001) and first 15 months of life (OR = 3.94, p < 0.0001) than controls, whereas cases diagnosed with febrile seizure were no more likely than controls to have received increased organic Hg. On a per microgram of organic Hg basis, cases diagnosed with a PDD in comparison to controls were at significantly greater odds (OR = 1.0197, p < 0.0001) of receiving increasing organic Hg exposure within the first 15 months of life, whereas cases diagnosed febrile seizure were no more likely than controls (OR = 0.999, p > 0.20) to have received increasing organic Hg exposure within the first 15 months of life. Routine childhood vaccination is an important public health tool to reduce the morbidity and mortality associated with infectious diseases, but the present study provides new epidemiological evidence of a significant relationship between increasing organic Hg exposure from Thimerosal-containing vaccines and the subsequent risk of PDD diagnosis in males and females.
Sigma-Aldrich
Thimerosal, 97.0-101.0% (on dried substance, titration)
Thimerosal, European Pharmacopoeia (EP) Reference Standard
USP
Thimerosal, United States Pharmacopeia (USP) Reference Standard